missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-fluoro-4-nitrobenzene, 95%, Thermo Scientific Chemicals

Product Code. 15401079
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-45-1
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD00055433
InChI Key FAWMTDSAMOCUAR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoronitrobenzene, 1-bromo-2-fluoro-5-nitrobenzene, 3-bromo-4-fluoro-1-nitrobenzene, 2-bromo-1-fluoro-4-nitro-benzene, 2-bromo-4-nitrofluorobenzene, benzene, 2-bromo-1-fluoro-4-nitro, pubchem8499, acmc-1birt, 3-bromo-4-fluoronitrobezene, ksc494c7t
PubChem CID 69699
IUPAC Name 2-bromo-1-fluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)F

Specifications

Quantity 100 g
Beilstein 2089367
Solubility Information Slightly soluble in water.
Formula Weight 220
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-1-fluoro-4-nitrobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'