missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-benzofuran, 97%, Thermo Scientific™

Product Code. 10212972
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10212972 10 g 10g
10592662 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10212972 Supplier Thermo Scientific Maybridge Supplier No. CC06610EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54008-77-4
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
InChI Key RNEOFIVNTNLSEH-UHFFFAOYSA-N
Synonym 2-bromobenzofuran, 2-bromobenzo b furan, bromobenzofuran, 2-bromo-1-benzofurane, benzofuran, 2-bromo, pubchem13460, 2-bromanyl-1-benzofuran, 2-bromo-1-benzo b furan
PubChem CID 2776264
IUPAC Name 2-bromo-1-benzofuran
SMILES C1=CC=C2C(=C1)C=C(O2)Br

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 197.03
Chemical Name or Material 2-Bromo-1-benzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.