missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(5-chloro-3-methylbenzo[b]thiophen-2-yl)ethan-1-one, 95%, Thermo Scientific™

Product Code. 10396944
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10396944 10 g 10g
10722123 250 mg 250mg
10029361 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10396944 Supplier Thermo Scientific Maybridge Supplier No. MWP00203EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-97-1
Molecular Formula C11H8BrClOS
Molecular Weight (g/mol) 303.60
MDL Number MFCD00173792
InChI Key HBNULIMIUQEVFS-UHFFFAOYSA-N
Synonym 2-bromo-1-5-chloro-3-methylbenzo b thiophen-2-yl ethan-1-one, 2-bromo-1-5-chloro-3-methyl-1-benzothiophen-2-yl ethanone, 2-bromo-1-5-chloro-3-methylbenzo b thiophen-2-yl ethanone, 2-bromoacetyl-5-chloro-3-methylbenzo b thiophene, 2-bromo-1-5-chloro-3-methyl-1-benzothien-2-yl ethanone, ethanone,2-bromo-1-5-chloro-3-methylbenzo b thien-2-yl, 2-bromanyl-1-5-chloranyl-3-methyl-1-benzothiophen-2-yl ethanone, 2-bromo-1-5-chloro-3-methyl-1-benzothiophen-2-yl ethan-1-one, 2-bromo-1-5-chloro-3-methyl-benzo b thiophen-2-yl ethan-1-one
PubChem CID 2779873
SMILES CC1=C(SC2=CC=C(Cl)C=C12)C(=O)CBr

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
IUPAC Name 2-bromo-1-(5-chloro-3-methyl-1-benzothiophen-2-yl)ethanone
Formula Weight 303.61
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(5-chloro-3-methylbenzo[b]thiophen-2-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.