missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone, Thermo Scientific™

Product Code. p-3075390
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10560683

Brand: Thermo Scientific Maybridge MO07142DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-70-3
Molecular Formula C10H8Br2O2
Molecular Weight (g/mol) 319.98
MDL Number MFCD06200889
InChI Key NPGVDFRSCHXENE-UHFFFAOYSA-N
Synonym 2-bromo-1-5-bromo-2,3-dihydro-1-benzofuran-7-yl ethanone, 2-bromo-1-5-bromo-2,3-dihydrobenzofuran-7-yl ethanone, 2-bromo-1-5-bromo-2,3-dihydro-1-benzo b furan-7-yl ethanone, 2-bromo-1-5-bromo-2,3-dihydro-1-benzofuran-7-yl ethan-1-one
PubChem CID 2794800
IUPAC Name 2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone
SMILES C1COC2=C1C=C(C=C2C(=O)CBr)Br

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 319.98
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(5-bromo-2, 3-dihydro-1-benzofuran-7-yl)ethanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.