missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone, Thermo Scientific™

Product Code. 10275844
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10275844 5 g 5g
10560683 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275844 Supplier Thermo Scientific Maybridge Supplier No. MO07142DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-70-3
Molecular Formula C10H8Br2O2
Molecular Weight (g/mol) 319.98
MDL Number MFCD06200889
InChI Key NPGVDFRSCHXENE-UHFFFAOYSA-N
Synonym 2-bromo-1-5-bromo-2,3-dihydro-1-benzofuran-7-yl ethanone, 2-bromo-1-5-bromo-2,3-dihydrobenzofuran-7-yl ethanone, 2-bromo-1-5-bromo-2,3-dihydro-1-benzo b furan-7-yl ethanone, 2-bromo-1-5-bromo-2,3-dihydro-1-benzofuran-7-yl ethan-1-one
PubChem CID 2794800
IUPAC Name 2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone
SMILES C1COC2=C1C=C(C=C2C(=O)CBr)Br

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 319.98
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(5-bromo-2, 3-dihydro-1-benzofuran-7-yl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.