missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-phenoxyphenyl)ethanone 95+%, Thermo Scientific™

Product Code. 10032464
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 28179-33-1
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.14
MDL Number MFCD01419855
InChI Key RAXTYMXDSNWNJS-UHFFFAOYSA-N
Synonym 2-bromo-1-4-phenoxyphenyl ethanone, 2-bromo-1-4-phenoxyphenyl ethan-1-one, ethanone,2-bromo-1-4-phenoxyphenyl, 4-phenoxyphenacyl bromide, 2-bromo-4'-phenoxyacetophenone, 4'-phenoxy-2-bromoacetophenone, alpha-bromo-4'-phenoxyacetophenone, ethanone, 2-bromo-1-4-phenoxyphenyl
PubChem CID 14797675
IUPAC Name 2-bromo-1-(4-phenoxyphenyl)ethanone
SMILES BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Assay Percent Range 90%
Quantity 250 mg
Formula Weight 291.14
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-phenoxyphenyl)ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'