missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-methylphenyl)ethanone, Mikromol™

Product Code. 30049336
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30049336 25 mg 25mg
30051045 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 30049336

Brand: Mikromol MM0333.130025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Bromo-1-(4-methylphenyl)ethanone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-4'-methylacetophenone
CAS 619-41-0
Analyte or Component Names 2-Bromo-4'-methylacetophenone
Shipping Condition Room Temperature
Molecular Formula C9 H9 Br O
Certifications/Compliance ISO 9001
Synonym Ethanone, 2-bromo-1-(4-methylphenyl)-, Acetophenone, 2-bromo-4'-methyl- (6CI,7CI,8CI), 2-Bromo-1-(4-methylphenyl)ethanone, 2-Bromo-1-(4-methylphenyl)ethan-1-one, 2-Bromo-1-(4-tolyl)ethanone, 2-Bromo-1-(p-methylphenyl)ethanone, 2-Bromo-1-(p-tolyl)ethanone, 2-Bromo-4'-methylacetophenone, 2-Bromo-p-methylacetophenone, 4-Methylphenacyl bromide, 4'-Methyl-2-bromoacetophenone, Bromomethyl 4-methylphenyl ketone, Bromomethyl p-tolyl ketone, NSC 63192, p-Methyl-Ω-bromoacetophenone, p-Methylphenacyl bromide, Ω-Bromo-p-methylacetophenone, 4-Methyl-alpha-bromoacetophenone, p-Methyl-alpha-bromoacetophenone, p-Methyl-omega-bromoacetophenone, alpha-Bromo-4'-methylacetophenone, alpha-Bromo-p-methylacetophenone, omega-Bromo-p-methylacetophenone
InChI Formula InChI=1S/C9H9BrO/c1-7-2-4-8(5-3-7)9(11)6-10/h2-5H,6H2,1H3
SMILES Cc1ccc(cc1)C(=O)CBr
IUPAC Name 2-bromo-1-(4-methylphenyl)ethanone
Molecular Weight (g/mol) 213.07
Quantity 25 mg
Formula Weight 211.9837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.