missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-methylphenyl)ethan-1-one, 97%, Maybridge™

Product Code. p-2697844
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
50g
Product Code. Quantity Packaging unitSize
10386474 10 g Amber Glass Bottle 10g
10396474 50 g Amber Glass Bottle 50g
2 options
This item is not returnable. View return policy

Product Code. 10386474

Brand: Thermo Scientific Maybridge CD00282EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 619-41-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
InChI Key KRVGXFREOJHJAX-UHFFFAOYSA-N
Synonym 2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
PubChem CID 69272
IUPAC Name 2-bromo-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3C6H4COCH2Br
Quantity 10 g
Formula Weight 213.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(4-methylphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.