missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-{4-methyl-2-[4-(trifluoromethyl)phenyl]1,3-thiazol-5-yl}-1-ethanone, ≥97%, Thermo Scientific™

Product Code. 10092242
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10092242 1 g 1g
10799913 250 mg 250mg
10654205 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092242 Supplier Thermo Scientific Maybridge Supplier No. MO00858DA
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-81-6
Molecular Formula C13H9BrF3NOS
Molecular Weight (g/mol) 364.18
MDL Number MFCD05664414
InChI Key XGHUMHHAMHEPCS-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazol-5-yl ethanone, 2-bromo-1-4-methyl-2-4-trifluoromethyl phenyl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-4-trifluoromethyl phenyl-5-thiazolyl
PubChem CID 2794631
IUPAC Name 2-bromo-1-[4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl]ethanone
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)C(=O)CBr

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 364.18
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-{4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazol-5-yl}-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.