missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-methoxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10356524
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356524 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10356524 Supplier Thermo Scientific Maybridge Supplier No. CD00356EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2632-13-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H9BrO2
Linear Formula CH3OC6H4COCH2Br
MDL Number MFCD00000201
Quantity 10 g
Synonym 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
InChI Key XQJAHBHCLXUGEP-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C(=O)CBr
IUPAC Name 2-bromo-1-(4-methoxyphenyl)ethan-1-one
Molecular Weight (g/mol) 229.07
PubChem CID 4965
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(4-methoxyphenyl)ethan-1-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.