missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,4-dimethoxybenzene, 98%

Product Code. 11427547
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427547 250 g 250g
11407547 10 g 10g
11417547 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427547 Supplier Thermo Scientific Alfa Aesar Supplier No. A15640.30
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25245-34-5 | C8H9BrO2 | 217.06 g/mol

2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25245-34-5
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00008355
InChI Key DWCGNRKFLRLWCJ-UHFFFAOYSA-N
PubChem CID 91353
IUPAC Name 2-bromo-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(Br)=C1

Specifications

Density 1.441
Boiling Point 260°C to 262°C
Flash Point >110°C (230°F)
Refractive Index 1.57
Quantity 250 g
Beilstein 2441884
Synonym 1-bromo-2,5-dimethoxybenzene, benzene, 2-bromo-1,4-dimethoxy, 2,5-dimethoxybromobenzene, pubchem19785, 2-bromo-4-methoxyanisole, acmc-1cby3, ksc490m9p, 1,4-dimethoxy-2-bromobenzene, 1-bromo-2, 5-dimethoxybenzene, dwcgnrkflrlwcj-uhfffaoysa
Solubility Information Insoluble in water.
Formula Weight 217.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,4-dimethoxybenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.