missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one, Technical Grade, Thermo Scientific™

Product Code. 10334192
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10334192 10 g Amber Glass Bottle 10g
10427622 1 g Amber Glass Bottle 1g
10327014 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10334192 Supplier Thermo Scientific Maybridge Supplier No. JP00700EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22019-49-4
Molecular Formula C8H5BrClNO3
Molecular Weight (g/mol) 278.486
InChI Key FXNQTMRPLLNPQY-UHFFFAOYSA-N
PubChem CID 2778330
IUPAC Name 2-bromo-1-(4-chloro-3-nitrophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)[N+](=O)[O-])Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 2-bromo-1-4-chloro-3-nitrophenyl ethanone, 2-bromo-1-4-chloro-3-nitrophenyl ethan-1-one, 4-chloro-3-nitrophenacylbromide, 4-chloro-3-nitrophenacyl bromide, ethanone,2-bromo-1-4-chloro-3-nitrophenyl, 4-chloro-3-nitro-phenacyl bromide, 4-chloro-3-nitrophen-acyl bromide, 2-bromo-4'-chloro-3'-nitroacetophenone, 2-bromo-1-4-chloro-3-nitro-phenyl ethanone, ethanone, 2-bromo-1-4-chloro-3-nitrophenyl
Formula Weight 278.49
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.