missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-thiazole-4-carboxylic acid, 98%

Product Code. 15204378 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15204378

Brand: Thermo Scientific Acros 454600050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5198-88-9 | C4H2BrNO2S | 208.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5198-88-9
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.03
InChI Key BEGREHRAUWCAHV-UHFFFAOYSA-N
Synonym 2-bromo-4-thiazolecarboxylic acid, 2-bromothiazole-4-carboxylic acid, 2-bromo-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-bromo, 2-bromo-4-thiazolecarboxylicacid, 2-bromothiazole-4-carboxylicacid, 2-bromo-4-carboxy-1,3-thiazole, pubchem22900, acmc-1avkr, 2-bromo-4-carboxythiazole
PubChem CID 2763209
IUPAC Name 2-bromo-1,3-thiazole-4-carboxylic acid
SMILES C1=C(N=C(S1)Br)C(=O)O

Specifications

Melting Point 204.0°C to 205.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 208.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-thiazole-4-carboxylic acid
compliance-icons
Product Identifier
  • 2-Bromo-1,3-thiazole-4-carboxylic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.