missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one 95%, Thermo Scientific™

Product Code. 10089481
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089481 1 g Amber Glass Bottle 1g
10062264 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10089481 Supplier Thermo Scientific Maybridge Supplier No. SPB04811DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14731-14-7
Molecular Formula C11H8BrNO2
Molecular Weight (g/mol) 266.09
MDL Number MFCD00173898
InChI Key XTBXGZOVSCTNEC-UHFFFAOYSA-N
Synonym 5-bromoacetyl-3-phenylisoxazole, 2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one, 2-bromo-1-3-phenylisoxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one, 2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone, ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl, pubchem8717, buttpark 43\57-72
PubChem CID 2735532
IUPAC Name 2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
SMILES BrCC(=O)C1=CC(=NO1)C1=CC=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 266.09
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.