missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene, 97%

Product Code. 11394656
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394656 1 g 1g
11384656 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394656 Supplier Thermo Scientific Alfa Aesar Supplier No. H32580.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118754-55-5 | C7H2BrCl2F3O | 309.891 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118754-55-5
Molecular Formula C7H2BrCl2F3O
Molecular Weight (g/mol) 309.891
MDL Number MFCD03412210
InChI Key CAAJZKZQEJSRJO-UHFFFAOYSA-N
Synonym 2-bromo-1,3-dichloro-5-trifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy bromobenzene, 4-bromo-3,5-dichloro-1-trifluoromethoxy benzene, benzene,2-bromo-1,3-dichloro-5-trifluoromethoxy, acmc-1c5si
PubChem CID 18947696
IUPAC Name 2-bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C(=C1Cl)Br)Cl)OC(F)(F)F

Specifications

Quantity 1 g
Formula Weight 309.9
Percent Purity 97%
Chemical Name or Material 2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.