missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one, Tech., Thermo Scientific™

Product Code. 10222492
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10222492

Brand: Thermo Scientific Maybridge BTB09098DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-93-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.543
MDL Number MFCD00067862
InChI Key RGMFWCVOMGKIJM-UHFFFAOYSA-N
Synonym 2-bromo-1-3-chloro-4-methylphenyl propan-1-one, 2-bromo-3'-chloro-4'-methylpropiophenone, 1-propanone,2-bromo-1-3-chloro-4-methylphenyl, 2-bromo-3'-chloro-4'methyl propiophenone, 2-bromo-1-3-chloro-4-methylphenyl-1-propanone, 2-bromanyl-1-3-chloranyl-4-methyl-phenyl propan-1-one
PubChem CID 2774950
IUPAC Name 2-bromo-1-(3-chloro-4-methylphenyl)propan-1-one
SMILES CC1=C(C=C(C=C1)C(=O)C(C)Br)Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 261.55
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.