missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-triisopropylbenzene, 96%

Product Code. 11437647
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437647 25 g 25g
11447647 100 g 100g
11457647 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437647 Supplier Thermo Scientific Alfa Aesar Supplier No. A15692.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 21524-34-5 | C15H23Br | 283.253 g/mol

2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21524-34-5
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.253
MDL Number MFCD00051547
InChI Key FUMMYHVKFAHQST-UHFFFAOYSA-N
Synonym 2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf
PubChem CID 140846
IUPAC Name 2-bromo-1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C

Specifications

Boiling Point 125°C to 126°C (4 mmHg)
Refractive Index 1.5225
Quantity 25 g
Beilstein 1953440
Solubility Information Slightly miscible with water.
Formula Weight 283.25
Percent Purity 96%
Chemical Name or Material 2-Bromo-1,3,5-triisopropylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.