missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trichlorobenzene, 97%

Product Code. 11491587
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11491587

Brand: Thermo Scientific Alfa Aesar B25000.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19393-96-5 | C6H2BrCl3 | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19393-96-5
Molecular Formula C6H2BrCl3
Molecular Weight (g/mol) 260.34
MDL Number MFCD00060653
InChI Key BAPPAFMEUDJAQI-UHFFFAOYSA-N
PubChem CID 140526
IUPAC Name 2-bromo-1,3,5-trichlorobenzene
SMILES ClC1=CC(Cl)=C(Br)C(Cl)=C1

Specifications

Quantity 25 g
Synonym 1-bromo-2,4,6-trichlorobenzene, 2,4,6-trichlorobromobenzene, benzene, 2-bromo-1,3,5-trichloro, benzene,2-bromo-1,3,5-trichloro, acmc-20aoxf, 1,3,5-trichloro-2-bromobenzene, 2,4,6-trichloro-1-bromobenzene, 2-bromo-1,3,5-trichloro-benzene, #, 2-bromanyl-1,3,5-tris chloranyl benzene
Formula Weight 260.34
Percent Purity 97%
Chemical Name or Material 2-Bromo-1,3,5-trichlorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.