missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[3,5-di(trifluoromethyl)phenyl]ethan-1-one, 95%, Thermo Scientific™

Product Code. p-2697907
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10693033

Brand: Thermo Scientific Maybridge SPB07614DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 131805-94-2
Molecular Formula C10H5BrF6O
Molecular Weight (g/mol) 335.043
MDL Number MFCD00792434
InChI Key ZEKBFXJTIAEUOF-UHFFFAOYSA-N
Synonym 1-3,5-bis trifluoromethyl phenyl-2-bromoethanone, 3',5'-bis trifluoromethyl-2-bromoacetophenone, 1-3,5-bis trifluoromethyl phenyl-2-bromoethan-1-one, 3,5-bistrifluoromethyl-phenacyl bromide, 3,5-bis trifluoromethyl phenacyl bromide, ethanone,1-3,5-bis trifluoromethyl phenyl-2-bromo, 3,5-bis trifluoromethyl phenacylbromide, a-bromo-3',5'-bis trifluoromethyl acetophenone, 2-bromo-1-3,5-bis trifluoromethyl phenyl ethanone
PubChem CID 2736170
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]-2-bromoethanone
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)CBr

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula (CF3)2C6H3C(O)CH2Br
Quantity 1 g
Formula Weight 335.04
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-[3, 5-di(trifluoromethyl)phenyl]ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.