missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[3-(4-chlorophenyl)-5-isoxazolyl]-1-ethanone, 97%, Thermo Scientific™

Product Code. 10223142
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10223142 250 mg 250mg
10170702 1 g 1g
10301645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10223142 Supplier Thermo Scientific Maybridge Supplier No. SPB05675CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 258506-49-9
Molecular Formula C11H7BrClNO2
Molecular Weight (g/mol) 300.536
MDL Number MFCD00662759
InChI Key OUHGJKFYGYXITB-UHFFFAOYSA-N
Synonym 2-bromo-1-3-4-chlorophenyl-5-isoxazolyl-1-ethanone, 5-bromoacetyl-3-4-chlorophenyl isoxazole, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethan-1-one, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl-isoxazol-5-yl ethanone, 2-bromo-1-3-4-chloro-phenyl-isoxazol-5-yl-ethanone, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethan-1-one, pubchem8718
PubChem CID 2799661
IUPAC Name 2-bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethanone
SMILES C1=CC(=CC=C1C2=NOC(=C2)C(=O)CBr)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 300.54
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-[3-(4-chlorophenyl)-5-isoxazolyl]-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.