missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, Thermo Scientific™

Product Code. 10422074
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422074 25 g Amber Glass Bottle 25g
10130692 1 g Amber Glass Bottle 1g
10293022 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10422074 Supplier Thermo Scientific Maybridge Supplier No. CD00248EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 26346-85-0
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.101
InChI Key GSCCVWPVPFIRJP-UHFFFAOYSA-N
Synonym 2-bromo-1-2,4-dimethylphenyl ethanone, 2-bromo-1-2,4-dimethylphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethylphenyl, 2-bromo-2',4'-dimethylacetophenone, 2,4-dimethylphenacyl bromide, 2,4-dimethyl phenacyl bromide, pubchem16796, 2,4-dimethylphenacyl bromid, ksc557o1l, alpha-bromo-2',4'-dimethylacetophenone
PubChem CID 2063450
IUPAC Name 2-bromo-1-(2,4-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C(=O)CBr)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)2C6H3C(O)CH2Br
Quantity 25 g
Formula Weight 227.1
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.