missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one, Thermo Scientific™

Product Code. 10408002
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10408002 1 g 1g
10581692 250 mg 250mg
10123444 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10408002 Supplier Thermo Scientific Maybridge Supplier No. CC01305DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4629-54-3
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.08
MDL Number MFCD00099402
InChI Key CSSHRKYOZTZFCX-UHFFFAOYSA-N
Synonym 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone, 1,4-benzodioxan-6-ylbromomethyl ketone, 2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone, ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl
PubChem CID 2776171
SMILES BrCC(=O)C1=CC=C2OCCOC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
Formula Weight 257.08
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(2, 3-dihydro-1, 4-benzodioxin-6-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.