missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)pyridine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11404490
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404490 1 g 1g
11494480 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404490 Supplier Thermo Scientific Alfa Aesar Supplier No. H50075.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 199296-40-7 | Molecular Formula: C11H14N2O3 | Molecular Weight: 222.244 g/mol

2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

Solubility
Soluble in water.

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199296-40-7
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.244
MDL Number MFCD08064228
InChI Key WZROBBWIJBBWQP-UHFFFAOYSA-N
Synonym tert-butyl 5-formylpyridin-2-yl carbamate, tert-butyl 5-formylpyridin-2-ylcarbamate, tert-butyl n-5-formylpyridin-2-yl carbamate, 2-boc-amino pyridine-5-carboxaldehyde, 5-formyl-pyridin-2-yl-carbamic acid tert-butyl ester, 5-formylpyridin-2-yl carbamic acid tert butyl ester, 2-boc-aminopyridine-5-aldehyde, 2-tert-butoxycarbonylamino pyridine-5-carboxaldehyde, carbamicacid, n-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 6-aminonicotinaldehyde, 6-boc protected
PubChem CID 22034247
IUPAC Name tert-butyl N-(5-formylpyridin-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)C=O

Specifications

Quantity 1 g
Sensitivity Air and light sensitive
Solubility Information Soluble in water.
Formula Weight 222.25
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)pyridine-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.