missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)pyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Product Code. 11484520
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484520 1 g 1g
11474520 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484520 Supplier Thermo Scientific Alfa Aesar Supplier No. H50094.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Heat and light sensitive. Incompatible with oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072944-99-0
Molecular Formula C16H25BN2O4
Molecular Weight (g/mol) 320.196
MDL Number MFCD08063072
InChI Key FYKIZSWVZIHFLF-UHFFFAOYSA-N
Synonym 2-boc-amino pyridine-3-boronic acid pinacol ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-3-boronic acid pinacol ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-n-boc-amino pyridine-3-boronic acid, pinacol ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridyl carbamate, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-carbamic acid tert-butyl ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate
PubChem CID 45925650
IUPAC Name tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)NC(=O)OC(C)(C)C

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 320.2
Percent Purity 98%
Chemical Name or Material 2-(Boc-amino)pyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.