missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)ethyl bromide, 96%

Product Code. 15481378
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481378 5 g 5g
15491378 250 mg 250mg
15471378 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481378 Supplier Thermo Scientific Alfa Aesar Supplier No. H55211.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39684-80-5 | C7H14BrNO2 | 224.10 g/mol

N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store at -20°C. Reactive with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39684-80-5
Molecular Formula C7H14BrNO2
Molecular Weight (g/mol) 224.10
MDL Number MFCD02683428
InChI Key TZRQZPMQUXEZMC-UHFFFAOYSA-N
Synonym tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide
PubChem CID 4103526
IUPAC Name tert-butyl N-(2-bromoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCBr

Specifications

Melting Point 30°C to 32°C
Linear Formula BrCH2CH2NHCO2C(CH3)3
Beilstein 2325117
Solubility Information Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
Quantity 5 g
Formula Weight 224.1
Percent Purity 96%
Chemical Name or Material 2-(Boc-amino)ethyl bromide
compliance-icons
Product Identifier
  • 2-(Boc-amino)ethyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: N-(2-Bromoethyl)carbamic acid tert-butylester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.