missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)-5-pyridinemethanol, 97%

Product Code. 11444490
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11444490

Brand: Thermo Scientific Alfa Aesar H50077.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 169280-83-5 | C11H16N2O3 | 224.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169280-83-5
Molecular Formula C11H16N2O3
Molecular Weight (g/mol) 224.26
MDL Number MFCD07186264
InChI Key BORDVYKLAFQRSP-UHFFFAOYSA-N
Synonym tert-butyl 5-hydroxymethyl pyridin-2-yl carbamate, 2-boc-amino-5-pyridinemethanol, tert-butyl 5-hydroxymethyl pyridin-2-ylcarbamate, tert-butyl n-5-hydroxymethyl pyridin-2-yl carbamate, 5-hydroxymethyl-pyridin-2-yl-carbamic acid tert-butyl ester, carbamic acid, 5-hydroxymethyl-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-20ahdn, 2-boc-amino-5-hydroxymethylpyridine, 6-tert-butyloxycarbonylamino-3-pyridylmethanol
PubChem CID 3575389
IUPAC Name tert-butyl N-[5-(hydroxymethyl)pyridin-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)CO

Specifications

Quantity 5 g
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)-5-pyridinemethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.