missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)-5-bromothiazole, 97%, Thermo Scientific™

Product Code. 15488408
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488408 1 g 1g
15498408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488408 Supplier Thermo Scientific Alfa Aesar Supplier No. H61828.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(Boc-amino)-5-bromothiazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405939-39-1
Molecular Formula C8H11BrN2O2S
Molecular Weight (g/mol) 279.152
MDL Number MFCD07368614
InChI Key OIBKBVFFZYCBAQ-UHFFFAOYSA-N
Synonym n-boc-2-amino-5-bromothiazole, tert-butyl 5-bromothiazol-2-yl carbamate, tert-butyl 5-bromothiazol-2-ylcarbamate, tert-butyl n-5-bromo-1,3-thiazol-2-yl carbamate, 5-bromo-thiazol-2-yl-carbamic acid tert-butyl ester, tert-butyl 5-bromo-1,3-thiazol-2-yl carbamate, carbamic acid, 5-bromo-2-thiazolyl-, 1,1-dimethylethyl ester, pubchem8907, n-boc-2-amno-5-bromothazole, tert-butyl-5-bromothiazol-2-ylcarbamate
PubChem CID 21300388
IUPAC Name tert-butyl N-(5-bromo-1,3-thiazol-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(S1)Br

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 279.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(Boc-amino)-5-bromothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.