missing translation for 'onlineSavingsMsg'
Learn More

2-(Bismethyl)aminomethylcyclohexanone Hydrochloride, TRC

Product Code. 30134403
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134403 25 g 25g
17924335 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134403 Supplier Toronto Research Chemicals Supplier No. TRCB58544525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42036-65-7
Molecular Formula C9 H17 N O . Cl H
Molecular Weight (g/mol) 191.7
Synonym (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride, Tramadol Hydrochloride Imp. E (EP) as Hydrochloride
IUPAC Name 2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
SMILES Cl.CN(C)CC1CCCCC1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H17NO.ClH/c1-10(2)7-8-5-3-4-6-9(8)11;/h8H,3-7H2,1-2H3;1H
Formula Weight 191.1077
Chemical Name or Material (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.