missing translation for 'onlineSavingsMsg'
Learn More

2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde, 97%, Thermo Scientific™

Product Code. 15438136
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 669091-00-3
Molecular Formula C23H23OP
Molecular Weight (g/mol) 346.41
MDL Number MFCD15143662
InChI Key PQIAXRGPKDEYLW-UHFFFAOYSA-N
Synonym 2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde, acmc-20ao6m, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
PubChem CID 71310677
IUPAC Name 2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
SMILES CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=CC=C1C=O

Specifications

Melting Point 106°C to 110°C
Color Yellow
Quantity 100 mg
Sensitivity Air Sensitive
Formula Weight 346.4
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'