missing translation for 'onlineSavingsMsg'
Learn More

2-Biphenylcarboxylic acid, 98%

Product Code. 10713831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713831 25 g Glass bottle 25g
13478298 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713831 Supplier Thermo Scientific Acros Supplier No. 106270250
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 947-84-2 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947-84-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002463
InChI Key ILYSAKHOYBPSPC-UHFFFAOYSA-N
Synonym 2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
PubChem CID 70357
IUPAC Name 2-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White to Beige
Boiling Point 199.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5C6H4CO2H
Quantity 25 g
Beilstein 09,669
Solubility Information Solubility in water: insoluble.
Formula Weight 198.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Biphenylcarboxylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.