missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzylpyridine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
2-Benzylpyridine
Specifications
Specifications
| Chemical Name or Material | 2-Benzylpyridine |
| CAS | 101-82-6 |
| Analyte or Component Names | 2-Benzylpyridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H11 N |
| Certifications/Compliance | ISO 17025 |
| Synonym | Pheniramine Maleate Imp. A (EP), Pheniramine Imp. A (EP), 2-Benzylpyridine, Pheniramine Maleate Impurity A, Pheniramine Impurity A |
| InChI Formula | InChI=1S/C12H11N/c1-2-6-11(7-3-1)10-12-8-4-5-9-13-12/h1-9H,10H2 |
| SMILES | C(c1ccccc1)c2ccccn2 |
| IUPAC Name | 2-benzylpyridine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction