missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylpyridine, 98+%, Thermo Scientific Chemicals

Catalog Number 11463414
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
101-82-6
C12H11N
169.23
MFCD00006352
PCFUWBOSXMKGIP-UHFFFAOYSA-N
pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
7581
2-benzylpyridine
C(C1=CC=CC=C1)C1=CC=CC=N1
This item is not returnable. View return policy
101-82-6
C12H11N
169.23
MFCD00006352
PCFUWBOSXMKGIP-UHFFFAOYSA-N
pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
7581
2-benzylpyridine
C(C1=CC=CC=C1)C1=CC=CC=N1

2-Benzylpyridine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzylpyridine is used in organic synthesis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 10°C to 12°C
Density 1.054
Boiling Point 140°C to 143°C (11 mmHg)
Flash Point 125°C (257°F)
Refractive Index 1.579
Quantity 50 g
Beilstein 115875
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 169.23
Percent Purity ≥98%
Chemical Name or Material 2-Benzylpyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'