missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylpyridine, 98+%

Product Code. 11453414
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453414 1000 g 1000g
11463414 50 g 50g
11473414 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453414 Supplier Thermo Scientific Alfa Aesar Supplier No. A10662.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-82-6 | C12H11N | 169.23 g/mol

2-Benzylpyridine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzylpyridine is used in organic synthesis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-82-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006352
InChI Key PCFUWBOSXMKGIP-UHFFFAOYSA-N
Synonym pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
PubChem CID 7581
IUPAC Name 2-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 10°C to 12°C
Density 1.054
Boiling Point 140°C to 143°C (11 mmHg)
Flash Point 125°C (257°F)
Refractive Index 1.579
Quantity 1000 g
Beilstein 115875
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 169.23
Percent Purity ≥98%
Chemical Name or Material 2-Benzylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.