missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyethanol, 98%

Product Code. 10287230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10287230 5 g Glass bottle 5g
10509653 1 g Glass bottle 1g
10701365 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10287230 Supplier Thermo Scientific Acros Supplier No. 314650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-08-2 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-08-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002868
InChI Key CUZKCNWZBXLAJX-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
PubChem CID 12141
IUPAC Name 2-phenylmethoxyethanol
SMILES OCCOCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0700g/mL
Boiling Point 265°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2CH2OH
Refractive Index 1.5200 to 1.522
Quantity 5 g
Specific Gravity 1.07
Formula Weight 152.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Benzyloxyethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.