missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzaldehyde, 98%

Product Code. 11393416
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393416 50 g 50g
11383416 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393416 Supplier Thermo Scientific Alfa Aesar Supplier No. L12866.18
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5896-17-3 | C14H12O2 | 212.25 g/mol

2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5896-17-3
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00016583
InChI Key PBEJTRAJWCNHRS-UHFFFAOYSA-N
Synonym 2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde
PubChem CID 344784
IUPAC Name 2-phenylmethoxybenzaldehyde
SMILES O=CC1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 46°C to 47°C
Quantity 50 g
Beilstein 780307
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 2-Benzyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.