missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzaldehyde 98%, ACROS Organics™

Product Code. 10146713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10146713 10 g Glass bottle 10g
10473555 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10146713 Supplier Thermo Scientific Acros Supplier No. 392430100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5896-17-3
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00016583
InChI Key PBEJTRAJWCNHRS-UHFFFAOYSA-N
Synonym 2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde
PubChem CID 344784
IUPAC Name 2-(benzyloxy)benzaldehyde
SMILES O=CC1=CC=CC=C1OCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.15g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4C(=O)H
Refractive Index 1.5980 to 1.6020 (20°C, 589nm)
Quantity 10 g
Beilstein 08, III, 145
Formula Weight 212.25
Percent Purity ≥97.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-Benzyloxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.