missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Benzyloxyaniline, 97%

Product Code. 11436120
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436120 1 g 1g
11446120 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436120 Supplier Alfa Aesar™ Supplier No. H27065.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 20012-63-9
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD03093873
InChI Key PLPVLSBYYOWFKM-UHFFFAOYSA-N
Synonym 2-benzyloxyaniline, 2-benzyloxy aniline, benzenamine, 2-phenylmethoxy, 2-benzyloxyanilline, 2-benzyloxy-aniline, acmc-1cmch, 2-aminophenyl benzyl ether, 3-amino-4-benzyloxybenzene
PubChem CID 240548
IUPAC Name 2-phenylmethoxyaniline
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2N

Specifications

Melting Point 35°C to 39°C
Quantity 1 g
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material 2-Benzyloxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.