missing translation for 'onlineSavingsMsg'
Learn More

2-{[(Benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid, 97%, Maybridge™

Product Code. 10263612
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10263612 10 g 10g
11316945 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10263612 Supplier Thermo Scientific Maybridge Supplier No. SB02038EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1164-16-5
Molecular Formula C17H17NO5
Molecular Weight (g/mol) 315.33
MDL Number MFCD00037180
InChI Key MCRMUCXATQAAMN-GGYSOQFKNA-N
Synonym z-tyr-oh, n-cbz-l-tyrosine, n-benzyloxycarbonyl-l-tyrosine, cbz-l-tyrosine, n-carbobenzoxy-l-tyrosine, n-carbobenzyloxy-l-tyrosine, cbz-tyr-oh, benzyloxycarbonyl-l-tyrosine, n-cbz-l-tyrosine-oh, z-tyrosine
PubChem CID 712438
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5CH2OC(O)NHCH(CO2H)CH2C6H4OH
Quantity 10 g
Formula Weight 315.32
Percent Purity 97%
Chemical Name or Material 2-{[(Benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.