missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

£61.78 - £184.00

Chemical Identifiers

CAS 1661043-58-8
Synonym 2-Benzyloxy-3-fluorophenylboronic acid pinacol ester
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487136
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H62186.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15477136
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H62186.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1661043-58-8
2-Benzyloxy-3-fluorophenylboronic acid pinacol ester

Specifications

1661043-58-8
328.19
Solid
2-Benzyloxy-3-fluorophenylboronic acid pinacol ester
96%
2-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications