missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxy-2-methyl-1-propanol, 95%, Thermo Scientific Chemicals

Product Code. 11311919
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311919 1 g 1g
15443337 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311919 Supplier Thermo Scientific Alfa Aesar Supplier No. H26006.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91968-71-7 | Molecular Formula: C11H16O2 | Molecular Weight: 180.25 g/mol

2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91968-71-7
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
MDL Number MFCD08459314
InChI Key FRHJVAJQSPVSKV-UHFFFAOYSA-N
Synonym 2-benzyloxy-2-methylpropan-1-ol, 2-benzyloxy-2-methyl-1-propanol, 2-methyl-2-phenylmethoxy propan-1-ol
PubChem CID 42552903
IUPAC Name 2-methyl-2-phenylmethoxypropan-1-ol
SMILES CC(C)(CO)OCC1=CC=CC=C1

Specifications

Boiling Point 80°C to 85°C (0.1 mmHg)
Quantity 1 g
Solubility Information Immiscible with water.
Formula Weight 180.12
Percent Purity 95%
Chemical Name or Material 2-Benzyloxy-2-methyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.