missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)-1-ethanamine hydrochloride, ≥90%, Thermo Scientific™

Product Code. 10639724
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10639724 1 g Amber Glass Bottle 1g
10462813 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10639724 Supplier Thermo Scientific Maybridge Supplier No. MO07103DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10578-75-3
Molecular Formula C9H14ClNO
Molecular Weight (g/mol) 187.667
MDL Number MFCD06411550
InChI Key AFQMWBUNDONXED-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanamine hydrochloride, 2-benzyloxy-1-ethanamine hydrochloride, 2-benzyloxyethylamine hydrochloride, aminoethylbenzyl ether hydrochloride, 2-phenylmethoxy-ethylamine hydrochloride, 2-benzyloxy-1-ethanamine, hcl, 2-benzyloxy ethylamine hydrochloride, 2-benzyloxy ethyl amine hydrochloride, ethanamine, 2-phenylmethoxy-, hydrochloride, 2-phenylmethoxy ethylamine, chloride
PubChem CID 2794786
IUPAC Name 2-phenylmethoxyethanamine;hydrochloride
SMILES C1=CC=C(C=C1)COCCN.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 187.67
Percent Purity 97%
Chemical Name or Material 2-(Benzyloxy)-1-ethanamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.