missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Benzylmethylamino)-3',4'-dihydroxyacetophenone-d3, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2-(Benzylmethylamino)-3',4'-dihydroxyacetophenone-d3 |
| CAS | 1330172-65-0 |
| Molecular Formula | C16H14D3NO3 |
| Quantity | 100 mg |
| Synonym | 1-(3,4-Dihydroxyphenyl)-2-[methyl(phenylmethyl)amino]ethanone-d3, alpha-Benzylmethylamino-3,4-dihydroxyacetophenone-d3 |
| InChI Formula | InChI=1S/C16H17NO3/c1-17(10-12-5-3-2-4-6-12)11-16(20)13-7-8-14(18)15(19)9-13/h2-9,18-19H,10-11H2,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])N(CC(=O)c1ccc(O)c(O)c1)Cc2ccccc2 |
| IUPAC Name | 2-[benzyl(trideuteriomethyl)amino]-1-(3,4-dihydroxyphenyl)ethanone |
| Molecular Weight (g/mol) | 274.33 |
| Formula Weight | 274.1397 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction