missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylcyclohexanone, 97%, ACROS Organics™

Product Code. 15244278 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15244278 5 g 5g
15234278 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15244278 Supplier Thermo Scientific Acros Supplier No. 451860050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 946-33-8
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
InChI Key CUYLPYBYCYQGCE-UHFFFAOYSA-N
PubChem CID 98945
IUPAC Name 2-benzylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)CC2=CC=CC=C2

Specifications

Density 1.0240g/mL
Boiling Point 103.0°C to 105.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Linear Formula C6H5CH2C6H9O
Quantity 5 g
Specific Gravity 1.024
Synonym 2-benzylcyclohexanone, cyclohexanone, 2-phenylmethyl, 2-benzyl cyclohexanone, 2-benzyl-cyclohexan-1-one, 2-phenylmethyl cyclohexanone, cyclohexanone,2-phenylmethyl, 2rs-2-phenylmethylcyclohexanone
Formula Weight 188.27
Percent Purity 97%
Chemical Name or Material 2-Benzylcyclohexanone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzylcyclohexan-1-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.