missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzylamino)-1,3-thiazole-5-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10561492
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 342405-23-6
Molecular Formula C11H10N2O2S
Molecular Weight (g/mol) 234.27
MDL Number MFCD03086200
InChI Key REVWBJBCAIBQGE-UHFFFAOYSA-N
Synonym 2-benzylamino-1,3-thiazole-5-carboxylic acid, maybridge3_004319, 2-benzylamino thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-phenylmethyl amino
PubChem CID 2779647
IUPAC Name 2-(benzylamino)-1,3-thiazole-5-carboxylic acid
SMILES OC(=O)C1=CN=C(NCC2=CC=CC=C2)S1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 234.28
Percent Purity 95%
Chemical Name or Material 2-(Benzylamino)-1, 3-thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'