missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-D-proline hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15448507
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448507 1 g 1g
15458507 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448507 Supplier Thermo Scientific Alfa Aesar Supplier No. H52019.03
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111130-04-1
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.72
MDL Number MFCD06659152
InChI Key SPHJFGQFCBZKRU-UHFFFAOYNA-N
Synonym s-2-benzylpyrrolidine-2-carboxylic acid hydrochloride, s-alpha-benzylproline hcl, 2-benzyl-d-proline hydrochloride, 2s-2-benzylpyrrolidine-2-carboxylic acid hydrochloride, d-proline,2-phenylmethyl, s-2-benzyl-d-proline hydrochloride, s-alpha-benzyl-proline hydrochloride, 2-benzyl-d-proline-hydrogen chloride 1/1
PubChem CID 45026885
SMILES Cl.OC(=O)C1(CC2=CC=CC=C2)CCCN1

Specifications

IUPAC Name (2S)-2-benzylpyrrolidine-2-carboxylic acid;hydrochloride
Quantity 1 g
Formula Weight 241.72
Optical Rotation +33° (c=1 in 1N HCl)
Percent Purity 95%
Chemical Name or Material 2-Benzyl-D-proline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.