missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11440387
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440387 25 g 25g
11450387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440387 Supplier Thermo Scientific Alfa Aesar Supplier No. B21764.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59-97-2 | Molecular Formula: C10H13ClN2 | Molecular Weight: 196.678 g/mol

An alpha adrenergic blocking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-97-2
Molecular Formula C10H13ClN2
Molecular Weight (g/mol) 196.678
MDL Number MFCD00012693
InChI Key RHTNTTODYGNRSP-UHFFFAOYSA-N
Synonym tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride
PubChem CID 6048
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC=C2.Cl

Specifications

Melting Point 174°C to 176°C
Quantity 25 g
Sensitivity Hygroscopic
Merck Index 14,9506
Formula Weight 196.68
Percent Purity 99%
Chemical Name or Material 2-Benzyl-2-imidazoline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.