missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylpyridine, TRC

Product Code. 30137423
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137423 100 g 100g
17932765 25 g 25g
17993665 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137423 Supplier Toronto Research Chemicals Supplier No. TRCB209570100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-02-1
Molecular Formula C12 H9 N O
Molecular Weight (g/mol) 183.21
Synonym Phenyl(pyridin-2-yl)methanone, Doxylamine Hydrogen Succinate Imp. D (EP), 2-Benzoylpyridine
IUPAC Name phenyl(pyridin-2-yl)methanone
SMILES O=C(c1ccccc1)c2ccccn2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C12H9NO/c14-12(10-6-2-1-3-7-10)11-8-4-5-9-13-11/h1-9H
Formula Weight 183.0684
Chemical Name or Material Phenyl(pyridin-2-yl)methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.