missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Benzoylpyridine, 99%

Product Code. 11499832
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499832 250 g 250g
11489832 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499832 Supplier Alfa Aesar™ Supplier No. A10857.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
The Schiff bases(2-Benzoylpyridine ) and copper(II) complexes display moderate antifungal activities. 2-Benzoylpyridine is an aromatic substituent which has selective antiproliferative effects, with the majority having greater antineoplastic activity. Reaction of 2-benzoylpyridine N(4)-phenyl thiosemicarbazone (HL) with a variety of copper salts yielded a series of copper(II) complexes of the type [CuLX]. Complexes of 2-benzoylpyridine thiosemicarbazone (H2Bz4DH) as well as of its N(4)-methyl (H2Bz4Me) and N(4)-phenyl (H2Bz4Ph) analogues with iron(II), nickel(II) and zinc(II) were characterized by a variety of spectroscopic techniques.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-02-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006300
InChI Key GCSHUYKULREZSJ-UHFFFAOYSA-N
Synonym 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone
PubChem CID 7038
IUPAC Name phenyl(pyridin-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=N2

Specifications

Melting Point 41°C to 45°C
Boiling Point 313°C
Flash Point 167°C (332°F)
Quantity 250 g
Beilstein 120283
Formula Weight 183.21
Percent Purity 99%
Chemical Name or Material 2-Benzoylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.