missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoxazolinone, MedChemExpress

Product Code. 16499376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499376 Supplier Medchem Express Supplier No. HYW015818
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Benzoxazolinone is an anti-leishmanial agent with an LC50 of 40 μg/mL against L. donovani. A building block in chemical synthesis. 1,3-Benzoxazol-2(3H)-one derivatives have antimicrobial activity against a selection of Gram-positive, Gram-negative bacteria and yeasts. Derivatives as anti-quorum sensing agent.

2-Benzoxazolinone is an anti-leishmanial agent with an LC50 of 40 μg/mL against L. donovani. A building block in chemical synthesis. 1,3-Benzoxazol-2(3H)-one derivatives have antimicrobial activity against a selection of Gram-positive, Gram-negative bacteria and yeasts. Derivatives as anti-quorum sensing agent.

Specifications

Chemical Name or Material 2-Benzoxazolinone
Physical Form Solid
CAS 59-49-4
Color Light Brown to Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H5NO2
For Use With (Application) COVID-19-immunoregulation
Synonym 2-Benzoxazolone 1,3-Benzoxazol-2(3H)-one 2-Hydroxybenzoxazole
Solubility Information DMSO : 100 mg/mL (740.08 mM; Need ultrasonic) ∣H2O : 3.85 mg/mL (28.49 mM; Need ultrasonic)
SMILES O=C1OC2=CC=CC=C2N1
Molecular Weight (g/mol) 135.12
Formula Weight 135.12
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.