missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothiazolethiol, TRC

Product Code. 30347955
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30347955

Brand: Toronto Research Chemicals TRCB20665010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Mercaptobenzothiazole
CAS 149-30-4
Molecular Formula C7H5NS2
Quantity 10 g
Synonym 2(3H)-Benzothiazolethione, 2-Benzothiazolethiol (8CI), 2-Benzothiazolinethione (6CI,7CI), 1,3-Benzothiazole-2-thiol, 1,3-Benzothiazole-2-thione, 2,3-Dihydrobenzothiazole-2-thione, 2-Benzothiazolyl mercaptan, 2-MBT, 2-Mercapto-1H-benzothiazole, 2-Mercaptobenzothiazole, 2-Mercaptobenzthiazole, 2-Sulfanylbenzothiazole, Accel M, Accelerator M, Aero 407, Aero Promoter 412, Benz-1,3-thiazolidine-2-thione, Benzo[d]thiazole-2-thiol, Benzothiazolethiol, Captax, Captax MBT, DT 402, Dermacid, Ekagom G, Kaptaks, Kaptax, M 2, M 2 (rust inhibitor), MBT, MBT (vulcanization accelerator), Mebetizol, Mebetizole, Mebithizol, Mercaptobenzothiazole, Mercaptobenzthiazole, Mertax, NSC 2041, Nocceler M, Nocceler M-P, Nonflex NB, Nuodeb 84, OCID, Oricel M, Perkacit MBT, Pneumax MBT, RU 3, Rotax, Royal MBT, Sanceler M, Sanceler M-G, Soxinol M, Thiotax, Vulkacit M, Vulkacit Mercapto, Vulkacit Mercapto MG/C, Vulkacit Mercapto/C, Vulkacit Mercapto/MG, Vulkafil ZN 94TT01, Wobezit M, accelerator MBT
InChI Formula InChI=1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)
SMILES Sc1nc2ccccc2s1
IUPAC Name 1,3-benzothiazole-2-thiol
Molecular Weight (g/mol) 167.2513
Formula Weight 166.9863
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.